Qualitative filter papers, grade 802, Whatman™
A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
- Highest retention among standard filters
- Slow flow rate
- Fine-particle retention
- For qualitative analysis
The filter is wet-strengthened and for normal qualitative application it will not introduce any significant impurities into the filtrate. However, it is not recommended for Kjeldahl nitrogen analysis.
- Catalog No:
- 515-2896
- 515-2895
- 515-2898
- WHAT5802-6698
- 515-2897
- WHAT5802-385
- WHAT5802-320
- Grado:
- 802
- 802
- 802
- 802
- 802
- 802
- 802
- Peso:
- 73 g/m²
- 73 g/m²
- 73 g/m²
- 73 g/m²
- 73 g/m²
- 73 g/m²
- 73 g/m²
- Spessore:
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- Dimensioni pori max.:
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- Dimensioni pori min.:
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- Portata:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
Frequently Bought Together