- Metodo chimico:Barbituric acid derivative
- Conf:100 Tests
- Descrizione:Cyanide test strips, white - violet
- Range di misura:0 - 30 mg/l CN⁻
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage color scale. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale
- Always ready to use: No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of cyanide in solutions. The test strips come with all necessary reagents, so that measurements can be started immediately. Within three minutes one gets a reliable result.
Cyanide is extremely poisonous. The lethal dose is 1 mg/kg weight. Careful control of the cyanide concentration is therefore essential, whenever cyanides are used for industrial processes like electroplating or gold-mining. Cyanide control is also important during the production of fruit-brandies made from stone fruits. These test strips are the ideal choice to check cyanide concentrations quickly and safely.
Gradation: 0 · 1 · 3 · 10 · 30 mg/l CN–