Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Catalog # QNTS081-3PTWH0XLID
Supplier: VWR Collection
Specifications
- Taglia dell'abbigliamento:XL
- Tipo di chiusura:Con chiusura a zip
- Applicazioni per abiti:Cleanroom
- Genere:Unisex
- Risk area:Anti-static
- Colore:bianco
- Seq:05
- Tessuto:Polyester/carbon
- Sleeve or leg length:Long
- Conf:1
Specifications
About this item
These coats have both zipper and pockets.
- Hanger in collar
- Stitched collar patch with labe
- Antistatic knitted cuffs
- One breast pocket and two hip pockets
- Two way zip in front
The coats contain press studs on left breast for PES clipband and also lap seams for tight and secure seam-enclosure.