Orders LinkContactUsLinkComponent
Detergent, Contec® NeutraKlean
Detergent, Contec® NeutraKlean
Nome del fornitore:   CONTEC
Detergent, Contec® NeutraKlean
Nome del fornitore:  CONTEC
Certificates
Detergent, Contec® NeutraKlean
Detergent, Contec® NeutraKlean
3 varianti
Product Details & Documents
Neutral detergent with purified water. NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms.
  • Neutral pH makes it suitable for use in all areas of cleanroom as gentle on surfaces
  • Filtered to 0.2 microns under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates
  • Low-foaming detergent action is ideal for use on floors, walls, and benches
  • Low level of surfactant provides low residue which can be easily removed
  • Trigger spray can be set to jet or spray and provides good surface coverage
  • Trigger spray and “bag in bottle” protected system prevents bottle contents from becoming contaminated during use
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Capped container is ideal for larger areas
Neutral pH after irradiation between 6,5 and 8,0 makes this as an ideal for carrying out the low-level cleaning required after maintenance, and to remove disinfectant residues and general soil.
The use of a surfactant/detergent helps to break the binding of particles and dirt to a surface and allows them to be more easily removed. Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life.
Certifications: The product is 0,2 μm filtered and filled under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates.
Informazioni per l'ordine: Ready to use product is available in a 1-gallon capped container, Also available as a irradiated trigger spray.
Informazioni per l'ordine: The product is supplied as either a 1 litre trigger spray or capped bottle and a 5 litre capped container for larger areas.
Confezione: Double bagged packed in linear tear packaging to aid the transfer of the product into a pharmaceutical cleanroom and also easy to open even when wearing gloves.
Product Family Options
Informazioni sul prodotto
  • Campo pH
    Confezione
    Disponibilità
    Price
  • CONTFBC103NK
    Contec® NeutraKlean
    Neutral detergent with purified water, capped
    Contec® NeutraKlean
    Neutral detergent with purified water, capped
    6,5 - 8,0
    Case of 6× 1 l capped bottles
    Log in per disponibilità
    Log in per disponibilità
    1 * 6 ST
    1 * 6 ST - SFr. 151.00
    SFr. 151.00
    1 * 6 ST
       SFr. 151.00
  • CONTFBC503NK
    Contec® NeutraKlean
    Neutral detergent with purified water, capped
    Contec® NeutraKlean
    Neutral detergent with purified water, capped
    6,5 - 8,0
    Case of 2× 5 l capped bottles
    Log in per disponibilità
    Log in per disponibilità
    1 * 2 ST
    1 * 2 ST - SFr. 251.00
    SFr. 251.00
    1 * 2 ST
       SFr. 251.00
  • CONTFBT103NK
    Contec® NeutraKlean
    Neutral detergent with purified water, spray
    Contec® NeutraKlean
    Neutral detergent with purified water, spray
    6,5 - 8,0
    Case of 6× 1 l trigger spray bottles
    Log in per disponibilità
    Log in per disponibilità
    1 * 6 ST
    1 * 6 ST - SFr. 239.00
    SFr. 239.00
    1 * 6 ST
       SFr. 239.00