Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Switzerland
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
Pagina principale
Standard
Standard per analisi ambientali
Nitrite, (CRM), Supelco®
Nitrite, (CRM), Supelco®
Nitrite, (CRM), Supelco®
Catalog # QC1153-2ML
Supplier:
Merck
CAS Number:
Expand image
Nitrite, (CRM), Supelco®
Catalog # QC1153-2ML
Supplier:
Merck
CAS Number:
Specifications
Conf:
2 mL
Descrizione:
Nitrite
Tipo:
Certified reference material
Specifications
About this item
Product Details
Product Details