Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Switzerland
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
Pagina principale
Standard
Standard per analisi ambientali
Chromium VI - Soil, (CRM), Supelco®
Chromium VI - Soil, (CRM), Supelco®
Chromium VI - Soil, (CRM), Supelco®
Catalog # CRM041-30G
Supplier:
Merck
CAS Number:
Certificates
Expand image
Chromium VI - Soil, (CRM), Supelco®
Catalog # CRM041-30G
Supplier:
Merck
CAS Number:
Certificates
Specifications
Conf:
30 g
Descrizione:
Chromium VI - Soil
Tipo:
Certified reference material
Specifications
About this item
Product Details
Documentation
Product Details
Documentation