Alkalinity calibration standards, (CRM), Supelco®
: Merck
Frequently Bought Together
VWR® AVS TITRINORM, Soluzioni tampone, 20 °C
: VWR Chemicals
HPLC waste-sets, V2.0
: S.C.A.T.
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Guanti in Nitrile
: VWR Collection
Dropping bulbs for pipettes and tubes
: KARTELL
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
EPA 8270 CALIBRATION MIX 500:1000MG/L 1 * 1 mL
: EHRENSTORFER
BOD seed inoculum, POLYSEED
: Hach
Acido borico ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. sostanza tampone, Fluka™
: Honeywell Chemicals