Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Home
Standard
Standard organici
Pharmaceutical secondary standard, rifaximin, (CRM), Supelco®
Pharmaceutical secondary standard, rifaximin, (CRM), Supelco®
Katalognr. PHR1946-300MG
Anbieter:
Merck
CAS-Nummer:
Bild vergrößern
Pharmaceutical secondary standard, rifaximin, (CRM), Supelco®
Katalognr. PHR1946-300MG
Anbieter:
Merck
CAS-Nummer:
Technische Daten
Conf:
300 mg
CAS No.:
80621-81-4
Descrizione:
RIFAXIMIN
Descrizione:
RIFAXIMIN
Technische Daten
Über diesen Artikel
Produktdetails
Produktdetails