Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standard
Standard organici
Pharmaceutical secondary standard, Glycerin, (CRM), Supelco®
Pharmaceutical secondary standard, Glycerin, (CRM), Supelco®
PHR1020-5G
:
Merck
:
Pharmaceutical secondary standard, Glycerin, (CRM), Supelco®
PHR1020-5G
:
Merck
:
Conf:
5 g
CAS No.:
56-81-5
Descrizione:
Glycerin
Descrizione:
Glycerin