SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standard
Standard organici
Pharmaceutical secondary standard, Ceftazidime, (CRM), Supelco®
Pharmaceutical secondary standard, Ceftazidime, (CRM), Supelco®
Pharmaceutical secondary standard, Ceftazidime, (CRM), Supelco®
Catalog # PHR1847-500MG
Supplier:
Merck
CAS Number:
Bild vergrößern
Pharmaceutical secondary standard, Ceftazidime, (CRM), Supelco®
Catalog # PHR1847-500MG
Supplier:
Merck
CAS Number:
Specifications
Conf:
500 mg
CAS No.:
78439-06-2
Descrizione:
Ceftazidime
Descrizione:
Ceftazidime
Specifications
About this item
Product Details
Product Details