Orders LinkContactUsLinkComponent
Columns, HiTrap™ Capto PlasmidSelect
Columns, HiTrap™ Capto PlasmidSelect
Nome del fornitore:   Cytiva
Columns, HiTrap™ Capto PlasmidSelect
Nome del fornitore:  Cytiva
Certificates
Columns, HiTrap™ Capto PlasmidSelect
2 varianti
Product Details & Documents
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • Bioprocess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.
Specifications for Product Family
Specifiche
  • Catalog No:
  • 17-5499-11
  • 17-5499-12
  • Pressione max.:
  • 5 bar
  • 5 bar
Specifiche
Product Family Options
Informazioni sul prodotto
  • Dimensione delle sfere
    Capacità
    Velocità del flusso
    Disponibilità
    Price
  • 17-5499-11
    HiTrap™ Capto PlasmidSelect
    HiTrap™ Capto PlasmidSelect
    7×25 mm
    1 ml
    <4 ml/min
    Log in per disponibilità
    Log in per disponibilità
    1 * 5 ST
    1 * 5 ST - SFr. 559.00
    SFr. 559.00
    1 * 5 ST
       SFr. 559.00
    Specifiche:
    More
    Specifications
    Cat. No.
    17-5499-11
    Pressione max.
    5 bar
  • 17-5499-12
    HiTrap™ Capto PlasmidSelect
    HiTrap™ Capto PlasmidSelect
    16×25 mm
    5 ml
    <20 ml/min
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 553.00
    SFr. 553.00
    1 * 1 ST
       SFr. 553.00
    Specifiche:
    More
    Specifications
    Cat. No.
    17-5499-12
    Pressione max.
    5 bar