Orders LinkContactUsLinkComponent
Multi-element noble metals standards, VHG
Multi-element noble metals standards, VHG
Certificates
Multi-element noble metals standards, VHG
2 Options Available Below
Product Details & Documents
Multi element standards for Au, Ir, Os, Pd, Pt, Re, Rh, Ru at 100 µg/ml in 20% HCl.
Product Family Options
Product Information
  • Descrizione
    Concentrazione
    Matrice
    Conf
    Availability
    Price
  • Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru
    100 mg/L
    20% HCl
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 477.00
    SFr. 477.00
    1 * 100 mL
       SFr. 477.00
  • Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru
    100 mg/L
    20% HCl
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 970.00
    SFr. 970.00
    1 * 500 mL
       SFr. 970.00