Cut resistant gloves, Showa S-TEX 581
Supplier: Showa
Certificates
About this item
These extremely lightweight hand-specific Hagane Coil®/Kevlar® gloves feature a microporeus foam nitrile coating, provide level 5-E cut resistance and naturally fits the anatomy of your hand for maximum precision.
- Nitrile coating provides excellent abrasion resistance and exceptional grip while allowing warm air and moisture to escape
- Hagane Coil® fibre reinforced with Kevlar® provides excellent cut resistance
- Embossed palm finish disperses oil away to increase grip and longevity in light oily environments
- Breathable back of hand to reduce perspiration
- Seamless knit to prevent irritation
- Suitable for contact heat resistance up to 250 °C
- Ergonomically designed for easy movement and continuous wear
Ideal for automotive, construction, glass, masonry, steel and metals industries. Typical applications include: Assembly, bottling, general maintenance, handling components with sharp edges, metal fabrication, metallurgy and stamping.
Certifications: EN 388:2016 (4X42E); meets ANSI Level 5; Cat. II
Packaging: 10 pairs per polybag.
Specifications
- Catalog No:
- 112-6014
- 112-6015
- 112-6016
- 112-6012
- 112-6013
- Modello:
- S-TEX 581
- S-TEX 581
- S-TEX 581
- S-TEX 581
- S-TEX 581
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Lattice/senza lattice:
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Materiale rivestimento interno:
- Hagane Coil®/Kevlar®
- Hagane Coil®/Kevlar®
- Hagane Coil®/Kevlar®
- Hagane Coil®/Kevlar®
- Hagane Coil®/Kevlar®
- Finitura:
- Rough
- Rough
- Rough
- Rough
- Rough
- Calibro:
- 13
- 13
- 13
- 13
- 13
- Tipo polsino:
- Elastico
- Elastico
- Elastico
- Elastico
- Elastico
- Norma EN:
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)