General purpose gloves, Showa B0500B
: Showa
These hand-specific nylon gloves with microporous PU foam coated palm and fingertips are designed for precision applications where dexterity, cleanliness and comfort are a priority.
- PU coating protects the hand from oils and abrasion while remaining elastic
- Seamless knit to prevent irritation
- Ideal colour for working in dirty environments
- Light, elastic low-lint, resistant to deformation and snug fit to maximize finger dexterity and tactility
- Permeable coating and breathable back of hand reduces perspiration and keeps hands dry
- Designed for easy movement and continuous wear thanks to renowned ergonomic shaped hand mould
Ideal for use in electronics, logistics, manufacturing, transport and automotive industry. Typical applications include: Assembly, delicate operation, mechanical inspection, precision work, product protection.
: EN 388:2016 (3131X); Cat. II.
- Catalog No:
- 112-5981
- 112-5977
- 112-5980
- 112-5979
- 112-5978
- Modello:
- B0500B
- B0500B
- B0500B
- B0500B
- B0500B
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Lattice/senza lattice:
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Materiale rivestimento interno:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finitura:
- Polyurethane palm coating
- Polyurethane palm coating
- Polyurethane palm coating
- Polyurethane palm coating
- Polyurethane palm coating
- Calibro:
- 13
- 13
- 13
- 13
- 13
- Tipo polsino:
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Norma EN:
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
Frequently Bought Together