VWR® Centrifuge Tubes, Conical Bottom, Flip Cap
Anbieter: VWR Collection
Zertifikate
Über diesen Artikel
Centrifuge tubes are ideal for use in centrifuging samples or storage in many research areas.
- Manufactured from high grade polypropylene
- Easy flip-open caps enable single-handed access
- Autoclavable at 121 °C, and freezable to −80 °C
- Sterilised by electron beam irradiation, DNase-free, RNase-free and non pyrogenic
- During the freezing process, it is better to use plastic rack
- During high-speed centrifugation, it is better to use suitable rotor adapters
Tubes feature black printed graduations and writing area can withstand chloroform.
Certifications: ISO 13485:2003 and ISO 9001:2008.
Packaging: Racked tubes are packed in environmentally responsible (fully recyclable) chipboard racks. Bagged tubes are packed in durable, re-sealed (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability.
Technische Daten
- Catalog No:
- 525-1048
- 525-1045
- 525-1046
- 525-1047
- Environmentally Preferable:
- Produzione a basso impatto
- Produzione a basso impatto
- Produzione a basso impatto
- Produzione a basso impatto
- Certificazione ambientale:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Technische Daten
Frequently Bought Together