Orders LinkContactUsLinkComponent
Graphic mats
Graphic mats
Nome del fornitore:   ERGOMAT
Graphic mats
Nome del fornitore:  ERGOMAT
Certificates
Graphic mats
2 varianti
Product Details & Documents
These graphic logo mats are extremely strong and durable while being easy to clean and broadly resistant to incidental liquid spills. These mats are promoting a brand and decorate a space with a functional, practical health and safety product that delivers years of relief and aesthetic enjoyment.
  • Free of silicone and latex
  • Moulded in durable, lightweight polyurethane
  • Can be customised with logos or other desired graphics
Available with ergonomic bubble profile or with moderately textured profile and smooth logo: Infinity bubble logo mat is designed to allow workers an anti-fatigue, ergonomic work station. Smooth logo mat offers a flat surface to accommodate pivoting workers and allow trolley accessibility.
Certifications: Rated to ISO Cleanroom Class 5 (Class 100).
Product Family Options
Informazioni sul prodotto
  • L×L
    Spessore
    Disponibilità
    Price
  • ERGOLB6090
    LogoMat, bubble
    LogoMat, bubble
    900×600 mm
    14,5 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - 145.00 SFr.
    145.00 SFr.
    1 * 1 ST
       145.00 SFr.
  • ERGOLS6090
    LogoMat, smooth
    LogoMat, smooth
    900×600 mm
    11 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - 121.00 SFr.
    121.00 SFr.
    1 * 1 ST
       121.00 SFr.