Orders LinkContactUsLinkComponent
Scoops, long handled, SteriWare®
Scoops, long handled, SteriWare®
Supplier:   Sampling Systems
Scoops, long handled, SteriWare®
Supplier:  Sampling Systems
Certificates
Scoops, long handled, SteriWare®
Scoops, long handled, SteriWare®
7 Options Available Below
Product Details & Documents
These SteriWare® long handled scoops are the ideal tool for reaching into deep containers. The extra long handle will help operators keep their hands out of the product.
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Available sterile (SAL 10-6 ) and non sterile
They are all manufactured and packed in a cleanroom to ensure cleanliness and reduce particulate contamination.
These single use scoops are ideal for use in the laboratory, production and warehouse.
Handle length: 300 mm
Certifications: Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials. BSE-/TSE-free. Full batch traceability.
Packaging: Individually bagged.
Product Family Options
Product Information
  • Capacità
    Lunghezza
    Larghezza
    Sterili
    Materiale
    Availability
    Price
  • 25 ml
    356 mm
    34 mm
    -
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 175.00
    SFr. 175.00
    1 * 50 ST
       SFr. 175.00
  • 25 ml
    356 mm
    34 mm
    +
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 251.00
    SFr. 251.00
    1 * 50 ST
       SFr. 251.00
  • 50 ml
    371 mm
    40 mm
    -
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 175.00
    SFr. 175.00
    1 * 50 ST
       SFr. 175.00
  • 50 ml
    371 mm
    40 mm
    +
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 251.00
    SFr. 251.00
    1 * 50 ST
       SFr. 251.00
  • 100 ml
    390 mm
    50 mm
    -
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 175.00
    SFr. 175.00
    1 * 50 ST
       SFr. 175.00
  • 100 ml
    390 mm
    50 mm
    +
    HDPE
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 251.00
    SFr. 251.00
    1 * 50 ST
       SFr. 251.00
  • 100 ml
    +
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 270.00
    SFr. 270.00
    1 * 50 ST
       SFr. 270.00