Orders LinkContactUsLinkComponent
Vector, pET-24(+) DNA
Vector, pET-24(+) DNA
Vector, pET-24(+) DNA, Novagen®, 10 µG
Codice catalogo 69772-3
Nome del fornitore:   Merck Millipore (Novagen)
Vector, pET-24(+) DNA
Codice catalogo 69772-3
Nome del fornitore:  Merck Millipore (Novagen)
Numero CAS:  
Vector, pET-24(+) DNA
Vector, pET-24(+) DNA, Novagen®, 10 µG
Numero CAS:  
Price
SFr. 314.00
Log in per disponibilità
1 * 10 µG
Product Details & Documents
pET-24(+) is a transcription vector designed for expression from bacterial translation signals carried within a cloned insert. It lacks the ribosome binding site and ATG start codon present on the pET translation vectors. A C-terminal His·Tag® sequence is available
Delivery information: pET vectors are supplied as purified plasmid DNA (10 µg) and an induction control strain (supplied as a glycerol stock).
Specifiche
Specifiche
  • Conf:
    10 µG
  • Descrizione:
    Vector, pET-24(+) DNA, Novagen®
Specifiche