Orders LinkContactUsLinkComponent
Vector, pET-24d(+) DNA
Vector, pET-24d(+) DNA
Vector, pET-24d(+) DNA, Novagen®, 10 µG
Codice catalogo 69752-3
Nome del fornitore:   Merck Millipore (Novagen)
Vector, pET-24d(+) DNA
Codice catalogo 69752-3
Nome del fornitore:  Merck Millipore (Novagen)
Numero CAS:  
Vector, pET-24d(+) DNA
Vector, pET-24d(+) DNA, Novagen®, 10 µG
Numero CAS:  
Price
300.00 SFr.
Log in per disponibilità
1 * 10 µG
Product Details & Documents
The pET-24a-d(+) vectors carry an N-terminal T7·Tag® sequence plus an optional C-terminal His·Tag® sequence.
Informazioni per l'ordine: The pET vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).
Specifiche
Specifiche
  • Conf:
    10 µG
  • Descrizione:
    Vector, pET-24d(+) DNA, Novagen®