Strip di tappi ultratrasparenti e pellicola adesiva per piastre per qPCR
Codice catalogo 732-0081
Nome del fornitore: Thermo Fisher Scientific
Specifiche
- Colore:trasparente
- Descrizione:Strip di tappi ultra trasparenti, 8 per striscia
- Seq:02
- Conf:120
Specifiche
Informazioni su questo articolo
PP flat 8-cap strips designed for qPCR and other fluorescence applications.
- Compatible with all Thermo Scientific™ 96-well PCR plates, as well as standard and low profile tube strips
- Specially formulated ultra clear polypropylene enhances signal detection by minimising autoflourescence and maximising transmission
- Highly polished flat surface provides an optically clear window to deliver maximum and consistent signal transmission, critical for accurate qPCR results