TLC spray box
TLC spray box
Codice catalogo S1509-5EA
Nome del fornitore: Merck
Vai a
Product Details & Documents
This TLC spray box may be used to spray reagent for TLC plates. Can be used in thin layer chromatography and HPTLC.
The reagent/chemical spraying on the TLC plates are generally carried out inside a fume hood or commercially available TLC spray cabinet with blower and exhaust hose. The TLC plate can be placed supported upright or on a paper inside the spray box. Spraying the box ensures complete removal of reagent mist.
Compatible with most Sigma sprays.
Specifiche
- Descrizione:TLC spray box