Septum forceps, Aesculap®
200 mm
N° de catalogue AESCOL474R
Fournisseur: Aesculap
Aller à
Product Details & Documents
Cartilage morseliser forceps with removeable guard, converts cartilage into a membranous-like structure, which permits respositioning of the septum to midline.
Spécifications
- Lunghezza:200 mm