Cuvettes for fluorescence measurements, QS/QG
Supplier: HELLMA
Certificates
About this item
These general-purpose cuvettes are mainly designed for fluorescence measurements in the UV/visible wavelength range.
- Rectangular form
- Classified as a macro cell
- Made of quartz suprasill/quartz glass
Designed with four optical windows for static sampling and hence this cuvette will fit all standard spectrophotometer cell holders and can be used with any beam height.
Frequently Bought Together