Ceramic D-Torch™
Assembly including quartz outer tube for OPTIMA™ ICP kit (8x00)
Catalog # PERCN0777840
Supplier: PerkinElmer
Jump to:
Product Details & Documents
The ceramic outer tube is of particular benefit for the analysis of wear metals in engine oils, where quartz outer tubes often suffer from short lifetime.
- Analysis of high-matrix solutions – Fusion samples, samples with high-dissolved solids, samples with high-acid content
- Demountable quart or ceramic outer tubes
Also beneficial for Si determinations, where quartz outer tubes may produce high background signals.
Can cause early devitrification or sample deposition when using quartz.
Specifications
- Descrizione:Assembly including quartz outer tube for OPTIMA™ ICP kit (8x00)
Accessories
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777052Ceramic outer tubeOPTIMA™ ICP kits (4300/5300/7300V)Log in for availability1 * 1 ST - SFr. 2’720.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777053Quartz outer tubeOPTIMA™ ICP kits (4300/5300/7300V)Log in for availability1 * 1 ST - SFr. 361.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777508DV D-Torch quartz including outer tubeOPTIMA™ ICP kits (2x00/4x00/5x00/7x00)Log in for availability1 * 1 ST - SFr. 1’350.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777509DV base and inner tubeOPTIMA™ ICP kits (2x00/4x00/5x00/7x00)Log in for availability1 * 1 ST - SFr. 893.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777510DV quartz outer tubeOPTIMA™ ICP kits (2x00/4x00/5x00/7x00)Log in for availability1 * 1 ST - SFr. 413.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
Product Information
- Catalog #DescrizionePerAvailabilityPrice
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0774070Torch adapter with auxiliary portOPTIMA™ ICP kits (4300/5300/7300V)Log in for availability1 * 1 ST - SFr. 945.00
Product Information
- Catalog #DescrizionePerAvailabilityPrice
- PERCN0777880Ceramic outer tubeOPTIMA™ ICP kit (8x00)Log in for availability1 * 1 ST - SFr. 3’770.00