Biopsy punch with plunger system
MEGRKAI-P101
: MEGRO
- Ø:1 mm
- Descrizione:Biopsy punch with plunger system, sterile
- Materiale:Acciaio inossidabile
- Conf:20
The integrated collection aid (plunger system) permits the fast and easy removal of the skin sample from the biopsy punch.
- Ribbed handle
- Stainless steel blade