Kontaflon 85 release agent
Kontaflon 85 aerosol, 200 ml
Catalog # 291-1245
Supplier: HM INDUSTRIESERVICE
Jump to:
Product Details & Documents
Grease-free sliding release aerosol agent based on PTFE.
- Constantly temperature resistant up to +260 °C
Applicable in plastics processing, vulcanizing molds and chains etc.
Specifications
- Capacità:200 ml
- Descrizione:Kontaflon 85 aerosol