Manual pulse counter
MOIN1023MW
: MOINEAU
- Range di misura:0 − 9999
- Descrizione:Manual pulse counter
- Materiale:Metallo
- Conf:1
Manual mechanical counter with four digit key in a robust metal housing.
- General reset by the rotary knob side
- Adding one each finger pressure
- Folding ring to attach it to your finger