Qualitative filter papers, grade MN 617 wa
Catalog # MANA530018
Supplier: MACHEREY-NAGEL
Specifications
- Grado:MN 617 wa
- Taglia:185 mm
- Conf:100
Specifications
About this item
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.