Orders LinkContactUsLinkComponent
HPLC columns, NUCLEOSHELL® Phenyl-Hexyl | Avantor
HPLC columns, NUCLEOSHELL® Phenyl-Hexyl
Supplier:   MACHEREY-NAGEL
HPLC columns, NUCLEOSHELL® Phenyl-Hexyl
Supplier:  MACHEREY-NAGEL
Certificates
HPLC columns, NUCLEOSHELL® Phenyl-Hexyl
14 Options Available Below
Product Details & Documents
NUCLEOSHELL® Phenyl-Hexyl based on core-shell particle technology for fast and efficient HPLC.
  • Efficient core-shell silica gel
  • Phenyl hexyl modification
  • Hydrophobic phase with alternative selectivity in comparison to classical C18 modifications
The retention mechanisms are determined by π-π and hydrophobic interactions. The phase is recommended for aromatic and unsaturated compounds such as phthalates, benzodiazepines and beta-lactam antibiotics.
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Specifications for Product Family
Specifications
  • Catalog No:
  • MANA763732.20
  • MANA763734.20
  • MANA763736.20
  • MANA763732.30
  • MANA763734.30
  • MANA763736.30
  • MANA763732.40
  • MANA763734.40
  • MANA763736.40
  • MANA763732.46
  • MANA763734.46
  • MANA763736.46
  • MANA763738.20
  • MANA763738.30
  • Carico di carbonio:
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • 4,5%
  • Con endcapping:
  • Range di pH:
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
  • 1 - 10
Specifications
Product Family Options
Analytical columns
Product Information
  • Formato della colonna
    Fase
    Dimensioni pori
    Dimensioni particelle
    Ø int.
    Lunghezza
    Availability
    Price
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    2,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763732.20
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    2,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 923.00
    SFr. 923.00
    1 * 1 ST
       SFr. 923.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763734.20
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 923.00
    SFr. 923.00
    1 * 1 ST
       SFr. 923.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763736.20
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    3,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763732.30
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    3,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763734.30
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    3,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763736.30
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763732.40
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763734.40
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763736.40
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 890.00
    SFr. 890.00
    1 * 1 ST
       SFr. 890.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763732.46
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’030.00
    SFr. 1’030.00
    1 * 1 ST
       SFr. 1’030.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763734.46
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Analitica
    Phenyl-Hexyl
    90 Å
    2,7 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’030.00
    SFr. 1’030.00
    1 * 1 ST
       SFr. 1’030.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763736.46
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
Guard columns
Product Information
  • Formato della colonna
    Fase
    Dimensioni pori
    Dimensioni particelle
    Ø int.
    Lunghezza
    Availability
    Price
  • Precolonna
    Phenyl-Hexyl
    90 Å
    2,7 µm
    2,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 179.00
    SFr. 179.00
    1 * 3 ST
       SFr. 179.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763738.20
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10
  • Precolonna
    Phenyl-Hexyl
    90 Å
    2,7 µm
    3,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 179.00
    SFr. 179.00
    1 * 3 ST
       SFr. 179.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763738.30
    Carico di carbonio
    4,5%
    Con endcapping
    Range di pH
    1 - 10