ContactUsLinkComponent
Fixed anchorage plate, Protecta, 3M™
Catalog # CPSDAM210
Supplier:  CAPITAL SAFETY
undefined
Fixed anchorage plate, Protecta, 3M™
Catalog # CPSDAM210
Supplier:  CAPITAL SAFETY
Specifications
  • Descrizione:
    Fixed anchorage plate
  • Lunghezza:
    53 mm
  • Conf:
    1
Specifications
About this item
Protecta® Fixed anchorage plate made from stainless steel.
  • 12 mm fixing
  • 3 mm thick
Certifications: EN 795, Class A
Delivery: Supplied without fixing.