ContactUsLinkComponent
Remote anchoring system, First-Man-Up™, DBI-SALA®, 3M™
Remote anchoring system, First-Man-Up™, DBI-SALA®, 3M™
Catalog # CPSD2100094
Supplier:  CAPITAL SAFETY
undefined
Remote anchoring system, First-Man-Up™, DBI-SALA®, 3M™
Catalog # CPSD2100094
Supplier:  CAPITAL SAFETY
Specifications
  • Descrizione:
    Telescopic rod
  • Lunghezza:
    1,80 - 7,80 m
  • Seq:
    01
  • Conf:
    1
Specifications
About this item
The telescopic rod connector allows to attach a rope life line above the user, enabling the user to ascend a pylon or to reach the top of a pole in complete safety.
  • Rope life line up to 8 m
Certifications: EN 795, Class B