Trigger2Trigger coil tether, DBI-SALA®, 3M™
44 mm (relaxed), 700 mm (streched)
Codice catalogo CPSD1500068
Nome del fornitore: CAPITAL SAFETY
Trigger2Trigger coil tether, DBI-SALA®, 3M™
Codice catalogo CPSD1500068
Nome del fornitore: CAPITAL SAFETY
Vai a
Product Details & Documents
Lightweight vinyl construction - ideal alternative to classic bungee tethers.
- Ultra-compact design
- Easy to clean
Ideal for use from wristband.
Specifiche
- Lunghezza:44 mm (relaxed), 700 mm (streched)
- Cat. No.:CPSD1500068
- Peso:2,3 kg