Cleanroom shoe cover, PP/CPE, BioClean Dual™
Catalog # NITRS-BDOS
Supplier: Nitritex
Specifications
- Sterili:+
- Applicazioni per abiti:Cleanroom
- Colore:bianco
- Lunghezza:400 mm
- Conf:150 PAIR
- Classe di cleanroom:10/ISO 4
Specifications
About this item
Disposable shoe covers made from BioClean Dual™, a low linting spunbonded non-woven polypropylene fabric with embossed cast polyethylene sole for a secure footing.
- Durable
- Heavy duty slip-resistant sole
- Latex-free elastic
Suitable for use in ISO Class 4 environment.
Certifications: Manufactured in a facility holding ISO 9001:2008 certification and processed in an NEBB certified ISO Class 4 environment.
Packaging: One pair per sealed inner PE bag, 15 inner bags per sealed outer bag, 10 outer bags per lined carton (150 pairs).