Orders LinkContactUsLinkComponent
Sampling spoons, SteriWare®
Sampling spoons, SteriWare®
Supplier:   Sampling Systems
Sampling spoons, SteriWare®
Supplier:  Sampling Systems
Certificates
Sampling spoons, SteriWare®
Sampling spoons, SteriWare®
8 Options Available Below
Product Details & Documents
SteriWare® Single use long handled spoons are ideal for reaching into large containers or difficult to reach places. It can be used straight from the bag and manufactured under cleanroom conditions so they can be used without costly cleaning.
  • Made from FDA Conforming HDPE
  • Pre-sterilised versions are available
  • Moulded and suitable for powders and liquids
  • Fully validated sterilisation procedure (SAL 10⁻⁶)
  • Full batch traceability
Certifications: FDA, EU 10/2011 and EC 1935/2004 conformity; BSE/TSE free.
Packaging: Individually bagged in a heat-sealed laminate bag.
Product Family Options
Straight spoons
Product Information
  • Descrizione
    Lunghezza
    Capacità
    Availability
    Price
  • Sampling spoons, sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 357.00
    SFr. 357.00
    1 * 100 ST
       SFr. 357.00
  • Sampling spoons, non sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 283.00
    SFr. 283.00
    1 * 100 ST
       SFr. 283.00
  • Sampling spoons, sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 360.00
    SFr. 360.00
    1 * 100 ST
       SFr. 360.00
  • Sampling spoons, non sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 285.00
    SFr. 285.00
    1 * 100 ST
       SFr. 285.00
Offset spoons
Product Information
  • Descrizione
    Lunghezza
    Capacità
    Availability
    Price
  • Sampling spoons, sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 355.00
    SFr. 355.00
    1 * 100 ST
       SFr. 355.00
  • Sampling spoons, non sterile
    300 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 306.00
    SFr. 306.00
    1 * 100 ST
       SFr. 306.00
  • Sampling spoons, sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 361.00
    SFr. 361.00
    1 * 100 ST
       SFr. 361.00
  • Sampling spoons, non sterile
    300 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 309.00
    SFr. 309.00
    1 * 100 ST
       SFr. 309.00