Battery powered engraving tool, SP Bel-Art
Codice catalogo BELAF441500005
Nome del fornitore: Bel-Art Products, a Part of SP
Specifiche
- Descrizione:Battery powered engraving tool
- Conf:1
Specifiche
Informazioni su questo articolo
Battery powered engraving tool to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
- Cordless battery power makes it portable for use virtually anywhere indoors or out in the field
- Marks will not fade if exposed to chemicals or sunlight
- Operates on two 'AA' batteries
Simply hold the engraving tool like a pen, press the power button, apply light pressure and write on test tubes, bottles, racks, keys, equipment, tools, electronics and more.
Ordering information: Convenient polyethylene stand (sold separately) with safe place to keep extra tip (tip included). AA batteries are not included.