Labels for MiniMark printer, type B-7569
Nome del fornitore: Brady
Vai a
Informazioni su questo articolo
The B-7569 vinyl labelling tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
- Gloss finish
- Permanent acrylic
19 varianti
Product Details & Documents
The B-7569 vinyl labelling tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
- Gloss finish
- Permanent acrylic
Product Family Options
Informazioni sul prodotto
- TipoModelloMaterialeAdesivoPrinting methodColoreStampanteLarghezzaDimensioniDisponibilitàPrice
- BRDY120853Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM BLACKVinileAcrilico permanenteThermal transferneroMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120851Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM BLUEVinileAcrilico permanenteThermal transferbluMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120850Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM GREENVinileAcrilico permanenteThermal transferverdeMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120852Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM ORANGEVinileAcrilico permanenteThermal transferarancioneMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120849Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM REDVinileAcrilico permanenteThermal transferrossoMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120847Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM WHITEVinileAcrilico permanenteThermal transferbiancoMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120848Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 29MM YELLOWVinileAcrilico permanenteThermal transfergialloMiniMark™29 mm29 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 270.00
- BRDY120860Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM BLACKVinileAcrilico permanenteThermal transferneroMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- BRDY120858Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM BLUEVinileAcrilico permanenteThermal transferbluMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- BRDY120859Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM ORANGEVinileAcrilico permanenteThermal transferarancioneMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- BRDY120856Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM REDVinileAcrilico permanenteThermal transferrossoMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- BRDY120854Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM WHITEVinileAcrilico permanenteThermal transferbiancoMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- BRDY120855Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 57MM YELLOWVinileAcrilico permanenteThermal transfergialloMiniMark™57 mm57 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 390.00
- 811-0186Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM BLACKVinileAcrilico permanenteThermal transferneroMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
- 811-0184Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM BLUEVinileAcrilico permanenteThermal transferbluMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
- 811-0185Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM ORANGEVinileAcrilico permanenteThermal transferarancioneMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
- 811-0182Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM REDVinileAcrilico permanenteThermal transferrossoMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
- 811-0180Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM WHITEVinileAcrilico permanenteThermal transferbiancoMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
- 811-0181Label printer tapeLabel printer tapeB-7569TAPE MINIMARK B7569 101MM YELLOWVinileAcrilico permanenteThermal transfergialloMiniMark™101 mm101 mm×35 mLog in per disponibilità1 * 1 ROUSFr. 585.00
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..