Orders LinkContactUsLinkComponent
Gas purification systems
Gas purification systems
Manifold and mounting hardware
Codice catalogo PERCN9306139
Nome del fornitore:   PerkinElmer
Gas purification systems
Codice catalogo PERCN9306139
Nome del fornitore:  PerkinElmer
Gas purification systems
Manifold and mounting hardware
Price
SFr. 1’130.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Moisture, oxygen, and other trace contaminants found in carrier or make-up gases can greatly affect your analysis and instrument performance. They can cause ghost peaks and degrade your column. Gas filters and traps can remove up to 99,9999% of these unwanted contaminants, increasing the life of your column life and decreasing stationary phase bleed.
  • Easy cartridge replacement with on/off knob
  • Double seal construction for safety
  • Two indicators for both oxygen and moisture
  • High capacity and efficiency levels for oxygen, water and hydrocarbons
Delivery information: Systems contain a manifold, mounting hardware and filter cartridge.
Specifiche
  • Descrizione:
    Manifold and mounting hardware
  • Seq:
    03