Thermal transfer printer labels, type B-423
Supplier: Brady
Jump to:
About this item
Polyester labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves
This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
27 Options Available Below
Product Details & Documents
Polyester labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves
This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Documents
TDS B-423 Labels and Signs TDS B-423Y Labels and Signs TDS Bradybondz B-423 Thermal Transfer Printable Glossy White Polyester Label Stock TDS Bradybondz B-423Y Thermal Transfer Printable Glossy Yellow Polyester Label Stock TDS Bradybondz™ B-423 thermal transfer printable glossy white polyester label stock
Datasheet
Product Family Options
Product Information
- DescrizioneTipoModelloMaterialeAdesivoPrinting methodColoreStampanteDimensioniConfezioneAvailabilityPrice
- Printer labelsB-423THT-12-423-10PoliestereAcrilico permanenteThermal transferbiancoi710012×11 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 178.00
- Printer labelsB-423THT-1-423-10PoliestereAcrilico permanenteThermal transferbiancoi510019×6 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 135.00
- Printer labelsB-423THT-41-423-10PoliestereAcrilico permanenteThermal transferbianco360X-Plus II25×4 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 215.00
- Printer labelsB-423THT-51-423-10PoliestereAcrilico permanenteThermal transferbiancoi510025×6 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 182.00
- Printer labelsB-423THT-29-423-10PoliestereAcrilico permanenteThermal transferbiancoi510031,7×9,5 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 247.00
- Printer labelsB-423THT-59-423-10PoliestereAcrilico permanenteThermal transferbianco360X-Plus II25×12 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 292.00
- Printer labelsB-423THT-5-423-10PoliestereAcrilico permanenteThermal transferbiancoi510025×12 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 292.00
- Printer labelsB-423THT-31-423-10PoliestereAcrilico permanenteThermal transferbianco360X-Plus II27×9 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 292.00
- Printer labelsB-423THT-43-423-10PoliestereAcrilico permanenteThermal transferbianco360X-Plus II31×6 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 204.00
- Printer labelsB-423THT-40-423-10PoliestereAcrilico permanenteThermal transferbiancoi510034×6 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 224.00
- Printer labelsB-423THT-45-423-10PoliestereAcrilico permanenteThermal transferbiancoi510038×6 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 224.00
- Printer labelsB-423THT-37-423-10PoliestereAcrilico permanenteThermal transferbiancoi510038×12 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 345.00
- Printer labelsB-423THT-15-423-2.5PoliestereAcrilico permanenteThermal transferbianco360X-Plus II50×6 mm2500 labels/rollLog in for availability1 * 1 ROUSFr. 98.20
- Printer labelsB-423THT-53-423-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II50×12 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 177.00
- Printer labelsB-423THT-17-423-3PoliestereAcrilico permanenteThermal transferbianco600X-Plus II50×25 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 267.00
- Printer labelsB-423THT-17-423-3-YLPoliestereAcrilico permanenteThermal transfergiallo600X-Plus II50×25 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 275.00
- Printer labelsB-423THT-142-423-2.5PoliestereAcrilico permanenteThermal transferbianco360X-Plus II63×5 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 268.00
- Printer labelsB-423THT-7-423-3PoliestereAcrilico permanenteThermal transferbiancoi510069×31 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 463.00
- Printer labelsB-423THT-24-423-1PoliestereAcrilico permanenteThermal transferbianco360X-Plus II69×44 mm1000 labels/rollLog in for availability1 * 1 ROUSFr. 1’060.00
- Printer labelsB-423THT-18-423-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II76×25 mm3000 labels/rollLog in for availability1 * 1 ROUSFr. 404.00
- Printer labelsB-423THT-19-423-1PoliestereAcrilico permanenteThermal transferbianco360X-Plus II76×50 mm1000 labels/rollLog in for availability1 * 1 ROUSFr. 269.00
- Printer labelsB-423THT-21-423PoliestereAcrilico permanenteThermal transferbiancoi510091×76 mm1 roll/rollLog in for availability1 * 1 ROUSFr. 452.00
- Printer labelsB-423THT-55-423-1PoliestereAcrilico permanenteThermal transferbianco360X-Plus II101×50 mm1000 labels/rollLog in for availability1 * 1 ROUSFr. 347.00
- Printer labelsB-423THT-79-423-10PoliestereAcrilico permanenteThermal transferbiancoi710050,8×10,16 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 400.00
- Printer labelsB-423THT-6-423-10PoliestereAcrilico permanentebiancoi710038.1×19.05 mmLog in for availability1 * 1 ROUSFr. 517.00
- Printer labelsB-423THT-103-423-10PoliestereAcrilico permanenteThermal transferbiancoi710025.4×6.35 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 211.00
- Printer labelsB-423THT-3-423-10PoliestereAcrilico permanentebiancoi710025.4×9.53 mm10000 labels/rollLog in for availability1 * 1 ROUSFr. 204.00