Equivalence, Column Coupler
Catalog # EQV-C0001
Supplier: Avantor
Specifications
- Descrizione:Column coupler
- Conf:1
Specifications
About this item
This column coupler is mainly used with guard column cartridges for narrow bore columns 1,0 mm ID and semi-prep cartridge holder.