Orders LinkContactUsLinkComponent
Rectangular cuvettes | Avantor
Rectangular cuvettes
 :   BIOCHROM
Rectangular cuvettes
 :  BIOCHROM
Rectangular cuvettes
Product Family Options
  • Descrizione
    Altezza
    Volume
    Cammino ottico
    Materiale finestra
  • Rectangular cell with lid
    45 mm
    200 µl
    1 mm
    Quarzo UV
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with lid
    45 mm
    200 µl
    1 mm
    Vetro ottico
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with lid
    45 mm
    1 ml
    5 mm
    Quarzo UV
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with stopper
    48 mm
    2 ml
    10 mm
    Quarzo UV
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with lid
    45 mm
    2 ml
    10 mm
    Vetro ottico
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with lid
    45 mm
    2,5 ml
    10 mm
    Quarzo UV
    1 * 1 ST
    1 * 1 ST
  • Rectangular cell with lid
    45 mm
    10 ml
    50 mm
    Quarzo UV