DNA transfection reagent, NeuroMag™
Supplier: OZ BIOSCIENCES
Certificates
About this item
NeuroMag™ transfection reagent is the first dedicated Magnetofection™ transfection reagent for neurons from 1 DIV to 21 DIV. It has proven to be extremely efficient in transfecting a large variety of primary neurons, such as cortical, hippocampal, dorsal root ganglion and motor neurons with all types of nucleic acids.
- Great efficiency, ideal for primary neurons
- Efficient from 1 DIV to 21 DIV
- Non toxic, high transfected neurons viability
- Ready to use, straightforward and rapid
- For all types of nucleic acids
High transfection efficiency has also been achieved in primary astrocytes, oligodendrocyte precursors or neural stem cells as well as other cell lines (C6, B65, PC12, N2A, etc.). Due to its unique properties, NeuroMag™ allows transfected neurons to be followed for several days.
Store at –20 °C.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery: Shipped at room temperature.
Specifications
- Specifico per cella:Neurons
- DNA:Yes
- mRNA:Yes
- Oligos:Yes
- shRNA:Yes
- siRNA:Yes