Magnetic plates
Nome del fornitore: OZ BIOSCIENCES
Vai a
Informazioni su questo articolo
Optimised magnetic plates with tailored properties especially designed for Magnetofection™.
- Can be easily cleaned and decontaminated with 70% ethanol
- Can be used at room temperature, 37 °C, +4 °C, etc.
- Compatible with culture plates from most common suppliers
- Solid, completely reusable
Suitable for all Magnetofection™ reagents, all cell culture dishes and flasks and can be used within incubators and with robots.
3 varianti
Product Details & Documents
Optimised magnetic plates with tailored properties especially designed for Magnetofection™.
- Can be easily cleaned and decontaminated with 70% ethanol
- Can be used at room temperature, 37 °C, +4 °C, etc.
- Compatible with culture plates from most common suppliers
- Solid, completely reusable
Suitable for all Magnetofection™ reagents, all cell culture dishes and flasks and can be used within incubators and with robots.
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- OZBIMF10000Super magnetic plate, 8×12 cm
- OZBIMF10096Magnetic plate, 96 magnets, 8×12 cm
- OZBIMF14000Mega magnetic plate, 20×26 cm