Attest™ 390 Auto-Reader for Steam
Catalog # MMMEM390
Supplier: 3M MEDICAL
Specifications
- Larghezza:229 mm
- Descrizione:Auto-reader for steam, 390
- Lunghezza:222 mm
- Conf:1
Specifications
About this item
This auto-reader for steam, 390, is designed to incubate and automatically read the biological indicators for steam at 60 °C for a final fluorescent result.
- Reliable and accurate
- Features advanced electronics and a backlit LCD panel
- Intelligent display with time-to-result countdown feature
- Audible alarm sounds for improved monitoring
- Display can be viewed remotely using your smartphone or PC
Colour: Light grey/charcoal