Sartorius wedge for windows
Catalog # SARBYSW02
Supplier: Sartorius Balances
Specifications
- Descrizione:Sartorius Wedge for Windows
- Conf:1
- Per:SART
Specifications
About this item
Sartorius wedge is a software package designed to capture data from aartorius devices into windows applications, specifically microsoft excel.
- Eliminate manual data entry errors
- Log data from multiple devices simultaneously
- Capture real-time data to any Windows application
- Parse and filter the data to match your requirements
- Accurate, reliable, and compliant
Sartorius wedge includes two of TALtech's data capture programs: WinWedge and TCPWedge. WinWedge connects to most serial or USB devices and TCPWedge connects to most networked devices.