Pellet presses
Pellet press, Ø: 1/8"
Codice catalogo PARR2814
Nome del fornitore: PARR
Vai a
Product Details & Documents
Pellet presses used for compressing powdered samples into pellet or tablet form for oxygen bomb calorimetry.
- Pellets produced in cylindrical shape with flat ends
Suitable for pressing powdered material into briquette form.
Specifiche
- Descrizione:Pellet press, Ø: 1/8"
- Seq:2