Adapters with external thread
Nome del fornitore: PerkinElmer
Vai a
Informazioni su questo articolo
Adapters with external screw thread (6,4 mm) for FIMS-100, FIMS-400, FIAS-100 or FIAS-400 flow injection system.
5 varianti
Product Details & Documents
Adapters with external screw thread (6,4 mm) for FIMS-100, FIMS-400, FIAS-100 or FIAS-400 flow injection system.
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- PERCB0507918Adapter K with 1,8 mm O.Ø nipple
- PERCB0507920Adapter L with 2,8 mm O.Ø nipple
- PERCB0507919Adapter M with 4,0 mm O.Ø nipple
- PERCB0507949Adapter N with 0,7 mm O.Ø Pt/lr capillary nipple
- PERCB0507921Screw plug