Suture, Dafilon (Aesculap®)
Dafilon black sutures, 15 cm
AESCG118110
: Aesculap
Dafilon is a non-absorbable nylon monofilament suture.
- Minimum tissue dragging
- Inert material with good tissue compatibility
- Excellent pliability
Suitable for skin closure in ophthalmic surgery, reconstructive surgery, and microsurgery.
- Descrizione:Dafilon black sutures
- Lunghezza:15 cm