Affinity chromatography columns, HiScreen™ IXSelect
HiScreen™ IXSelect, 4,7 ml
Codice catalogo GEHE17-3714-10
Nome del fornitore: Cytiva
Affinity chromatography columns, HiScreen™ IXSelect
Codice catalogo GEHE17-3714-10
Nome del fornitore: Cytiva
Vai a
Product Details & Documents
HiScreen™ IXSelect columns are prepacked with IXSelect affinity chromatography medium. These columns are an excellent choice for method optimisation and parameter screening.
- Easily connected in series to achieve 20 cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Ulteriori dettagli sul prodotto
Specifiche
- Capacità:4,7 ml
- Descrizione:HiScreen™ IXSelect