Orders LinkContactUsLinkComponent
Caps for autosampler vials
Caps for autosampler vials
Small caps, use with ND18 vials, 19,8×5,2 mm, Polyethylene-push on, Colour-21390374|Colour-21390348
  MICS97010-SC18
 :   MICROSOLV
Caps for autosampler vials
  MICS97010-SC18
 :  MICROSOLV
Caps for autosampler vials
Caps for autosampler vials
Small caps, use with ND18 vials, 19,8×5,2 mm, Polyethylene-push on, Colour-21390374|Colour-21390348
SFr. 11.20
1 * 100 ST
  • Tipo:
    Polyethylene-push on
  • Descrizione:
    Small caps, use with ND18 vials, 19,8×5,2 mm
  • Colore tappo:
    Colour-21390374|Colour-21390348