Recombinant protein G, peroxidase conjugated, Calbiochem®
Catalog # 539322-500
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Conf:500 µG
- Descrizione:Recombinant Protein G, peroxidase conjugated
Specifications
About this item
Purified recombinant Protein G conjugated to peroxidase can be used in direct immunoassay.
- Used in enzyme-linked immunosorbent assay and immunoblotting (Western blotting)
- Binds ≥99% to human IgG immobilised on a column